1,11-Dodecadiene ≥99.0%

Supplier: TCI America

D0283-5ML D0283-25ML
TCD0283-005MLEA 284.64 CAD
TCD0283-005ML TCD0283-025ML
1,11-Dodecadiene ≥99.0%
1,11-Dodecadiene

CAS Number: 5876-87-9
MDL Number: MFCD00048156
Molecular Formula: C12H22
Molecular Weight: 166.31
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Specific Gravity (20/20): 0.77

Formula: C₁₂H₂₂
Boiling Pt: 207 °C
Density: 0.77 (20 °C)
MDL Number: MFCD00048156
CAS Number: 5876-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b484c9a909ce24f1323d7059a358b187ed60c28&vid=9b484c9a909ce24f1323d7059a358b187ed60c28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR