1,2-Di(4-pyridyl)ethylene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0276-25G D0276-10G
TCD0276-25GEA 1017.52 CAD
TCD0276-25G TCD0276-010G
1,2-Di(4-pyridyl)ethylene ≥98.0% (by GC, titration analysis)
1,2-Di(4-pyridyl)ethylene

CAS Number: 13362-78-2
MDL Number: MFCD00006448
Molecular Formula: C12H10N2
Molecular Weight: 182.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 1,2-Bis(4-pyridyl)ethylene

Formula: C₁₂H₁₀N₂
Melting Pt: 151 °C
MDL Number: MFCD00006448
CAS Number: 1135-32-6
EINECS: 214-491-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR