Didodecylamine ≥97.0%

Supplier: TCI America
Warning

Synonyms: Dilaurylamine

D0267-5G D0267-25G
TCD0267-005GEA 169.28 CAD
TCD0267-005G TCD0267-025G
Didodecylamine ≥97.0%
Didodecylamine

CAS Number: 3007-31-6
MDL Number: MFCD00041918
Molecular Formula: C24H51N
Molecular Weight: 353.68
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 265
Melting point (°C): 51


Other Product Names / Synonyms: Dilaurylamine

Formula: C₂₄H₅₁N
Boiling Pt: 265 °C
Melting Pt: 51 °C
MDL Number: MFCD00041918
CAS Number: 3007-31-6
EINECS: 221-114-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91582d37506aecc475ef5a56120607fd9f612a33&vid=91582d37506aecc475ef5a56120607fd9f612a33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR