1,6-Diaminohexane dihydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,6-Hexanediamine dihydrochloride, Hexane-1,6-diamine dihydrochloride, Hexamethylenediamine dihydrochloride

D0096-500G D0096-25G
TCD0096-500GEA 529.3 CAD
TCD0096-500G TCD0096-025G
1,6-Diaminohexane dihydrochloride ≥98.0% (by titrimetric analysis)
1,6-Diaminohexane dihydrochloride

CAS Number: 6055-52-3
MDL Number: MFCD00012528
Molecular Formula: C6H16N2
Molecular Weight: 189.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 248


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Hexamethylenediamine Dihydrochloride, 1,6-Hexanediamine Dihydrochloride

Formula: C₆H₁₈Cl₂N₂
MW: 189.13 g/mol
Melting Pt: 248 °C
Density: 1.216 g/cm³ (20 °C)
MDL Number: MFCD00012528
CAS Number: 6055-52-3
EINECS: 227-977-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77f6dd95011439b0a23253335ea7272af6956824&vid=77f6dd95011439b0a23253335ea7272af6956824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR