2,7-Diaminofluorene ≥98.0%
Supplier: TCI America
Synonyms:
Fluorene-2,7-diamine, 2,7-Diamino-fluorene, Fluorene-2,7-diyldiamine
D0092-5G
D0092-25G
D0092-1G
TCD0092-005GEA
418.7
CAD
TCD0092-005G
TCD0092-025G
TCD0092-1G
2,7-Diaminofluorene ≥98.0%
2,7-Diaminofluorene
CAS Number: 525-64-4
MDL Number: MFCD00001128
Molecular Formula: C13H12N2
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 164
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bioscience, Stains & Dyes
Other Product Names / Synonyms: DAF
Formula:
C₁₃H₁₂N₂ MW: 196.25 g/mol Boiling Pt: 467 Melting Pt: 165 |
MDL Number:
MFCD00001128 CAS Number: 525-64-4 EINECS: 208-377-5 Merck Index: 13,04182 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...