1,4-Dihydronaphthalene ≥80.0%

Supplier: TCI America

D0088-5G D0088-1G
TCD0088-5GEA 390.26 CAD
TCD0088-5G TCD0088-001G
1,4-Dihydronaphthalene ≥80.0%
1,4-Dihydronaphthalene

CAS Number: 612-17-9
MDL Number: MFCD00058963
Molecular Formula: C10H10
Molecular Weight: 130.19
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: DELTA2-Dialin

Formula: C₁₀H₁₀
Boiling Pt: 212 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00058963
CAS Number: 612-17-9
EINECS: 210-297-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b6a7edbb885d71d84c96cb655ff153537a9688&vid=f9b6a7edbb885d71d84c96cb655ff153537a9688&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR