3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0%

Supplier: TCI America
Warning

Synonyms: DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate

D0078-5G D0078-25G
TCD0078-005GEA 153.37 CAD
TCD0078-005G TCD0078-025G
3,3'-Diaminobenzidine tetrahydrochloride hydrate ≥98.0%
3,3'-Diaminobenzidine tetrahydrochloride hydrate

CAS Number: 868272-85-9
MDL Number: MFCD08273058
Molecular Formula: C12H14N4
Molecular Weight: 360.10
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 280
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Biphenyls (for High-Performance Polymer Research), Bioscience, Stains & Dyes
Other Product Names / Synonyms: 3,3',4,4'-Tetraaminobiphenyl Tetrahydrochloride, DAB.4HCl

Formula: C₁₂H₁₄N₄·4HCl·H₂O
MW: 360.11 g/mol (anhy)
Melting Pt: 280 °C
Storage Temperature: Freezer
MDL Number: MFCD08273058
CAS Number: 868272-85-9
EINECS: 231-018-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3b44aae59a6abd52aa1e04c26e260395165e210&vid=d3b44aae59a6abd52aa1e04c26e260395165e210&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR