3,3'-Diaminobenzidine (DAB) ≥98.0%
Supplier: TCI America
Synonyms:
[1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine
CAS Number: 91-95-2
MDL Number: MFCD00007725
Molecular Formula: C12H14N4
Molecular Weight: 214.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 177
Flash Point (°C): 200
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,3',4,4'-Tetraaminobiphenyl
Formula:
C₁₂H₁₄N₄ MW: 214.27 g/mol Melting Pt: 177 °C Flash Pt: 200 °C |
MDL Number:
MFCD00007725 CAS Number: 91-95-2 EINECS: 202-110-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...