Decylbenzene ≥98.0%
Supplier: TCI America
D0034-5ML
D0034-25ML
TCD0034-005MLEA
77.42
CAD
TCD0034-005ML
TCD0034-025ML
Decylbenzene ≥98.0%
Decylbenzene
CAS Number: 104-72-3
MDL Number: MFCD00008955
Molecular Formula: C16H26
Molecular Weight: 218.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 298
Melting point (°C): -14
Flash Point (°C): 110
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 1-Phenyldecane
Formula:
C₁₆H₂₆ Boiling Pt: 298 °C Melting Pt: –14 °C Density: 0.86 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008955 CAS Number: 104-72-3 EINECS: 203-230-1 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...