4-Chloro-6-methoxypyrimidine ≥98.0%

Supplier: TCI America

C2369-5G C2369-1G
TCC2369-5GEA 298.62 CAD
TCC2369-5G TCC2369-1G
4-Chloro-6-methoxypyrimidine ≥98.0%
4-Chloro-6-methoxypyrimidine

CAS Number: 26452-81-3
MDL Number: MFCD06200211
Molecular Formula: C5H5ClN2O
Molecular Weight: 144.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 80
Freezing point (°C): 30

Formula: C₅H₅ClN₂O
MW: 144.5590 g/mol
Boiling Pt: 80 °C
MDL Number: MFCD06200211
CAS Number: 26452-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dc9db55b40eed5103f2db80e7115f2bb07f225a&vid=2dc9db55b40eed5103f2db80e7115f2bb07f225a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR