3-Chloro-2,4,5-trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C2261-25G
C2261-5G
TCC2261-25GEA
330.22
CAD
TCC2261-25G
TCC2261-5G
3-Chloro-2,4,5-trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-2,4,5-trifluorobenzoic acid
CAS Number: 101513-77-3
MDL Number: MFCD00153101
Molecular Formula: C7H2ClF3O2
Molecular Weight: 210.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 114
Formula:
C₇H₂ClF₃O₂ Melting Pt: 114 °C |
MDL Number:
MFCD00153101 CAS Number: 101513-77-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...