3-Chloro-5-fluoroanisole ≥96.0%

Supplier: TCI America
Warning

C2218-1G C2218-5G
TCC2218-1GEA 75.84 CAD
TCC2218-1G TCC2218-5G
3-Chloro-5-fluoroanisole ≥96.0%
3-Chloro-5-fluoroanisole

CAS Number: 202925-08-4
MDL Number: MFCD00070770
Molecular Formula: C7H6ClFO
Molecular Weight: 160.57
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 79
Specific Gravity (20/20): 1.27

Formula: C₇H₆ClFO
Boiling Pt: 189 °C
Density: 1.27 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00070770
CAS Number: 202925-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=411d8a373777219fb197283ec907e03d05f22fa9&vid=411d8a373777219fb197283ec907e03d05f22fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR