1-Chloro-2-fluoro-4-nitrobenzene ≥97.0%

Supplier: TCI America

Synonyms: 4-Chloro-3-fluoronitrobenzene

C2195-25G C2195-5G
TCC2195-25GEA 257.54 CAD
TCC2195-25G TCC2195-5G
1-Chloro-2-fluoro-4-nitrobenzene ≥97.0%
1-Chloro-2-fluoro-4-nitrobenzene

CAS Number: 350-31-2
MDL Number: MFCD00215833
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 61

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 116 °C
Melting Pt: 61 °C
MDL Number: MFCD00215833
CAS Number: 350-31-2
EINECS: 206-500-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR