2-Chloro-3-fluorobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2128-1G C2128-5G
TCC2128-1GEA 44.56 CAD
TCC2128-1G TCC2128-5G
2-Chloro-3-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
2-Chloro-3-fluorobenzoic acid

CAS Number: 102940-86-3
MDL Number: MFCD00973903
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 171

Formula: C₇H₄ClFO₂
Melting Pt: 171 °C
MDL Number: MFCD00973903
CAS Number: 102940-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b8fe5c116b2556e537bdff757b654c516ecd6d3&vid=0b8fe5c116b2556e537bdff757b654c516ecd6d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR