2-Chloro-4-methyl-5-nitropyridine ≥98.0%
Supplier: TCI America
C2021-1G
C2021-5G
TCC2021-1GEA
52.93
CAD
TCC2021-1G
TCC2021-5G
2-Chloro-4-methyl-5-nitropyridine ≥98.0%
2-Chloro-4-methyl-5-nitropyridine
CAS Number: 23056-33-9
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 91
Melting point (°C): 38
Flash Point (°C): 113
Other Product Names / Synonyms: 2-Chloro-5-nitro-4-picoline
Formula:
C₆H₅ClN₂O₂ Boiling Pt: 91 °C Melting Pt: 38 °C Flash Pt: 113 °C |
MDL Number:
MFCD00010688 CAS Number: 23056-33-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...