2-Chloro-3-methyl-5-nitropyridine ≥98.0%
Supplier: TCI America
C2020-5G
C2020-1G
TCC2020-5GEA
173.8
CAD
TCC2020-5G
TCC2020-1G
2-Chloro-3-methyl-5-nitropyridine ≥98.0%
2-Chloro-3-methyl-5-nitropyridine
CAS Number: 22280-56-4
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 49
Other Product Names / Synonyms: 2-Chloro-5-nitro-3-picoline
Formula:
C₆H₅ClN₂O₂ Boiling Pt: 145 °C Melting Pt: 49 °C |
MDL Number:
MFCD00173686 CAS Number: 22280-56-4 EINECS: 244-889-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...