2-Chloro-4-methyl-3-nitropyridine ≥98.0%

Supplier: TCI America

C2012-1G C2012-5G
TCC2012-1GEA 41.08 CAD
TCC2012-1G TCC2012-5G
2-Chloro-4-methyl-3-nitropyridine ≥98.0%
2-Chloro-4-methyl-3-nitropyridine

CAS Number: 23056-39-5
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 113


Other Product Names / Synonyms: 2-Chloro-3-nitro-4-picoline

Formula: C₆H₅ClN₂O₂
Melting Pt: 53 °C
Flash Pt: 113 °C
MDL Number: MFCD00012347
CAS Number: 23056-39-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e049d0faad8d284e1f448539f639a558de22ec1a&vid=e049d0faad8d284e1f448539f639a558de22ec1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR