2-Chloro-4-methylpyridine ≥98.0%

Supplier: TCI America
Warning

C1992-25G C1992-5G
TCC1992-25GEA 120.08 CAD
TCC1992-25G TCC1992-5G
2-Chloro-4-methylpyridine ≥98.0%
2-Chloro-4-methylpyridine

CAS Number: 3678-62-4
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 2-Chloro-4-picoline

Formula: C₆H₆ClN
Boiling Pt: 99 °C
Melting Pt: 115 °C
Density: 1.16 (20 °C)
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60b0549addb73236c178286ce059a29411d533e0&vid=60b0549addb73236c178286ce059a29411d533e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR