4-Chloroisatin ≥97.0%
Supplier: TCI America
Synonyms:
4-Chloroindoline-2,3-dione
C1870-25G
C1870-5G
TCC1870-25GEA
127.98
CAD
TCC1870-25G
TCC1870-5G
4-Chloroisatin ≥97.0%
4-Chloroisatin
CAS Number: 6344-05-4
Molecular Formula: C8H4ClNO2
Molecular Weight: 181.58
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 258
Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 4-Chloroindole-2,3-dione, 4-Chloro-2,3-indolinedione
Formula:
C₈H₄ClNO₂ Melting Pt: 258 °C |
MDL Number:
MFCD00129178 CAS Number: 6344-05-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...