1-Cyclohexylpiperazine ≥98.0%
Supplier: TCI America
C1783-25G
C1783-5G
TCC1783-25GEA
200.66
CAD
TCC1783-25G
TCC1783-5G
1-Cyclohexylpiperazine ≥98.0%
1-Cyclohexylpiperazine
CAS Number: 17766-28-8
MDL Number: MFCD00044809
Molecular Formula: C10H20N2
Molecular Weight: 168.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 36
Formula:
C₁₀H₂₀N₂ Boiling Pt: 256 °C (1013 hPa) Melting Pt: 36 °C |
MDL Number:
MFCD00044809 CAS Number: 17766-28-8 EINECS: 241-750-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...