2-Chloro-5-nitroanisole ≥98.0%
Supplier: TCI America
C1773-25G
C1773-5G
TCC1773-25GEA
148.52
CAD
TCC1773-25G
TCC1773-5G
2-Chloro-5-nitroanisole ≥98.0%
2-Chloro-5-nitroanisole
CAS Number: 1009-36-5
MDL Number: MFCD00079739
Molecular Formula: C7H6ClNO3
Molecular Weight: 187.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83
Other Product Names / Synonyms: 2-Methoxy-4-nitrochlorobenzene
Formula:
C₇H₆ClNO₃ Melting Pt: 83 °C |
MDL Number:
MFCD00079739 CAS Number: 1009-36-5 EINECS: 213-768-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...