3-Chloro-2-fluoroaniline ≥98.0%
Supplier: TCI America
C1659-5G
C1659-25G
TCC1659-005GEA
26.86
CAD
TCC1659-005G
TCC1659-025G
3-Chloro-2-fluoroaniline ≥98.0%
3-Chloro-2-fluoroaniline
CAS Number: 2106-04-9
MDL Number: MFCD00069415
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 110
Specific Gravity (20/20): 1.34
Formula:
C₆H₅ClFN Boiling Pt: 99 °C Density: 1.34 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00069415 CAS Number: 2106-04-9 EINECS: 218-283-6 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...