2-Chloro-6-methoxy-3-nitropyridine ≥98.0%

Supplier: TCI America
Warning

C1636-5G C1636-25G
TCC1636-005GEA 74.42 CAD
TCC1636-005G TCC1636-025G
2-Chloro-6-methoxy-3-nitropyridine ≥98.0%
2-Chloro-6-methoxy-3-nitropyridine

CAS Number: 38533-61-8
MDL Number: MFCD00130268
Molecular Formula: C6H5ClN2O3
Molecular Weight: 188.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 100

Formula: C₆H₅ClN₂O₃
MW: 188.57 g/mol
Flash Pt: 100 °C
MDL Number: MFCD00130268
CAS Number: 38533-61-8
EINECS: 253-989-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa1985fe28b6a88133f8fb8e1b4799db838a9c81&vid=aa1985fe28b6a88133f8fb8e1b4799db838a9c81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR