2-Chloro-6-methoxybenzothiazole ≥96.0%

Supplier: TCI America

C1464-5G C1464-1G
TCC1464-5GEA 312.84 CAD
TCC1464-5G TCC1464-001G
2-Chloro-6-methoxybenzothiazole ≥96.0%
2-Chloro-6-methoxybenzothiazole

CAS Number: 2605-14-3
MDL Number: MFCD00671510
Molecular Formula: C8H6ClNOS
Molecular Weight: 199.65
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 54

Formula: C₈H₆ClNOS
Boiling Pt: 133 °C
Melting Pt: 54 °C
MDL Number: MFCD00671510
CAS Number: 2605-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a432b9f952362c06b99d5209d02707be75a7a98f&vid=a432b9f952362c06b99d5209d02707be75a7a98f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR