2-Chloro-4,6-dimethoxypyrimidine ≥98.0%
Supplier: TCI America
C1433-25G
C1433-10G
TCC1433-25GEA
150.42
CAD
TCC1433-25G
TCC1433-010G
2-Chloro-4,6-dimethoxypyrimidine ≥98.0%
2-Chloro-4,6-dimethoxypyrimidine
CAS Number: 13223-25-1
MDL Number: MFCD00274530
Molecular Formula: C6H7ClN2O2
Molecular Weight: 174.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Formula:
C₆H₇ClN₂O₂ Melting Pt: 103 °C |
MDL Number:
MFCD00274530 CAS Number: 13223-25-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...