2-Chloro-4-nitroanisole ≥98.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-methoxy-5-nitrobenzene

C1352-5G C1352-25G
TCC1352-5GEA 94.8 CAD
TCC1352-5G TCC1352-025G
2-Chloro-4-nitroanisole ≥98.0%
2-Chloro-4-nitroanisole

CAS Number: 4920-79-0
MDL Number: MFCD00060681
Molecular Formula: C7H6ClNO3
Molecular Weight: 187.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: 3-Chloro-4-methoxynitrobenzene

Formula: C₇H₆ClNO₃
Melting Pt: 95 °C
MDL Number: MFCD00060681
CAS Number: 4920-79-0
EINECS: 225-547-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17954a48276fa46e4f36f97f06aaeb6cf8096187&vid=30186e30c4d484bc50c6c3e794e03c8dca1832af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR