Ethyl methyl carbonate ≥98.0%

Supplier: TCI America

Synonyms: Carbonic acid ethyl methyl ester

C1342-10ML C1342-25ML
TCC1342-010MLEA 90.06 CAD
TCC1342-010ML TCC1342-025ML
Ethyl methyl carbonate ≥98.0%
Ethyl methyl carbonate

CAS Number: 623-53-0
MDL Number: MFCD00191398
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Carbonic Acid Ethyl Methyl Ester

Formula: C₄H₈O₃
MW: 104.1045 g/mol
Boiling Pt: 107 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00191398
CAS Number: 623-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8282f6cef01bd25baf45f6bfa15edafb5833380&vid=e8282f6cef01bd25baf45f6bfa15edafb5833380&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR