6-Chloro-2-hexanone ≥97.0%

Supplier: TCI America

C1333-25ML C1333-250ML
TCC1333-025MLEA 282.66 CAD
TCC1333-025ML TCC1333-250ML
6-Chloro-2-hexanone ≥97.0%
6-Chloro-2-hexanone

CAS Number: 10226-30-9
MDL Number: MFCD00191625
Molecular Formula: C6H11ClO
Molecular Weight: 134.60
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 91
Specific Gravity (20/20): 1.03
Storage Temperature: <0°C


Other Product Names / Synonyms: 4-Chlorobutyl Methyl Ketone, 1-Chloro-5-hexanone

Formula: C₆H₁₁ClO
Boiling Pt: 79 °C
Density: 1.03 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00191625
CAS Number: 10226-30-9
EINECS: 233-546-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28dabfe87393378d6ab7af967e322077ba5fafa6&vid=32af0e3358dfb81a440dedd3177ca663ac2c538a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR