Benzyl chloroacetate ≥97.0%

Supplier: TCI America

C1265-250G C1265-25G
TCC1265-250GEA 1194.48 CAD
TCC1265-250G TCC1265-025G
Benzyl chloroacetate ≥97.0%
Benzyl chloroacetate

CAS Number: 140-18-1
MDL Number: MFCD00157062
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 113
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: Chloroacetic Acid Benzyl Ester

Formula: C₉H₉ClO₂
Boiling Pt: 132 °C
Density: 1.21 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00157062
CAS Number: 140-18-1
EINECS: 205-400-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b2c76a691362060f8a53c5ecb434fa496253f2c&vid=7b2c76a691362060f8a53c5ecb434fa496253f2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR