2-Chlorobenzophenone ≥98.0%

Supplier: TCI America

C1150-500G C1150-25G C1150-100G
TCC1150-500GEA 752.08 CAD
TCC1150-500G TCC1150-025G TCC1150-100G
2-Chlorobenzophenone ≥98.0%
2-Chlorobenzophenone

CAS Number: 5162-03-8
MDL Number: MFCD00000558
Molecular Formula: C13H9ClO
Molecular Weight: 216.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 46
Flash Point (°C): 113

Formula: C₁₃H₉ClO
Boiling Pt: 180 °C
Melting Pt: 46 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000558
CAS Number: 5162-03-8
EINECS: 225-936-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2789bc0f3395649c652adf80d4bc1e1e45fca8a8&vid=2789bc0f3395649c652adf80d4bc1e1e45fca8a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR