1-(2-Chlorophenyl)piperazine 98,0

Supplier: TCI America

C1102-25G C1102-5G
TCC1102-025GEA 306.52 CAD
TCC1102-025G 76219-666
1-(2-Chlorophenyl)piperazine 98,0
1-(2-Chlorophenyl)piperazine

CAS Number: 39512-50-0
MDL Number: MFCD00040727
Molecular Formula: C10H13ClN2
Molecular Weight: 196.68
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.18

Formula: C₁₀H₁₃ClN₂
MW: 196.68 g/mol
Boiling Pt: 135 °C
Density: 1.18 (20 °C)
CAS Number: 39512-50-0
EINECS: 254-480-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR