1-Chloroethylethyl carbonate ≥98.0%
Supplier: TCI America
C1056-500G
C1056-25G
C1056-100G
TCC1056-500GEA
962.22
CAD
TCC1056-500G
TCC1056-025G
76219-664
1-Chloroethylethyl carbonate ≥98.0%
1-Chloroethyl ethyl carbonate
CAS Number: 50893-36-2
MDL Number: MFCD00040523
Molecular Formula: C5H9ClO3
Molecular Weight: 152.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 62
Specific Gravity (20/20): 1.14
Other Product Names / Synonyms: Carbonic Acid 1-Chloroethyl Ethyl Ester, 1-Chlorodiethyl Carbonate
Formula:
C₅H₉ClO₃ Boiling Pt: 160 °C Density: 1.14 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00040523 CAS Number: 50893-36-2 EINECS: 256-832-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...