(Chloromethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C1009-5G C1009-25G
TCC1009-005GEA 104.28 CAD
TCC1009-005G TCC1009-025G
(Chloromethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(Chloromethyl)triphenylphosphonium chloride

CAS Number: 5293-84-5
MDL Number: MFCD00011867
Molecular Formula: C19H17Cl2P
Molecular Weight: 347.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₉H₁₇Cl₂P
MDL Number: MFCD00011867
CAS Number: 5293-84-5
EINECS: 226-139-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a507fab9db8954f609e6bf949712e0e966867007&vid=a507fab9db8954f609e6bf949712e0e966867007&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR