4-Chloro-2-nitrobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

C0964-5G C0964-25G
TCC0964-005GEA 414.75 CAD
TCC0964-005G TCC0964-25G
4-Chloro-2-nitrobenzaldehyde ≥98.0%
4-Chloro-2-nitrobenzaldehyde

CAS Number: 5551-11-1
MDL Number: MFCD00056104
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₇H₄ClNO₃
Melting Pt: 68 °C
MDL Number: MFCD00056104
CAS Number: 5551-11-1
EINECS: 226-915-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca22ae4392a872f1be0587b812da39b0166210e1&vid=1647fc23c8c3880dcb85a792292f00710324e729&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR