1-Chlorodocosane ≥98.0%

Supplier: TCI America

Synonyms: Docosyl chloride

C0775-25G C0775-5G
TCC0775-025GEA 461.04 CAD
TCC0775-025G 76219-656
1-Chlorodocosane ≥98.0%
1-Chlorodocosane

CAS Number: 42217-03-8
MDL Number: MFCD00039383
Molecular Formula: C22H45Cl
Molecular Weight: 345.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41


Other Product Names / Synonyms: Docosyl Chloride

Formula: C₂₂H₄₅Cl
Melting Pt: 41 °C
MDL Number: MFCD00039383
CAS Number: 42217-03-8
EINECS: 255-710-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=348613cdc03bd466c747b06cab1434513d95a2a9&vid=348613cdc03bd466c747b06cab1434513d95a2a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR