Butyl carbamate ≥98.0%

Supplier: TCI America

Synonyms: Carbamic acid butyl ester

C0613-25G C0613-500G
TCC0613-025GEA 98.04 CAD
TCC0613-025G TCC0613-500G
Butyl carbamate ≥98.0%
Butyl carbamate

CAS Number: 592-35-8
MDL Number: MFCD00007967
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 204
Melting point (°C): 54
Flash Point (°C): 108


Other Product Names / Synonyms: Carbamic Acid Butyl Ester

Formula: C₅H₁₁NO₂
Boiling Pt: 204 °C
Melting Pt: 54 °C
Flash Pt: 108 °C
MDL Number: MFCD00007967
CAS Number: 592-35-8
EINECS: 209-751-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43afcb6cb6b6dac2b8e3b89ee77fb54ee927e057&vid=43afcb6cb6b6dac2b8e3b89ee77fb54ee927e057&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR