4-Cyclohexylcyclohexanol ≥98.0%

Supplier: TCI America

C0497-25G C0497-5G
TCC0497-025GEA 468.31 CAD
TCC0497-025G TCC0497-5G
4-Cyclohexylcyclohexanol ≥98.0%
4-Cyclohexylcyclohexanol

CAS Number: 2433-14-9
MDL Number: MFCD00060071
Molecular Formula: C12H22O
Molecular Weight: 182.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₁₂H₂₂O
MDL Number: MFCD00060071
CAS Number: 2433-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8908469ea600aca405b87099e92df104f872e5ff&vid=439fbbd89753ad5dcda5deb3e4aa07509eae96bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR