Butyl cyanoacetate ≥98.0%
Supplier: TCI America
C0440-25G
C0440-500G
TCC0440-025GEA
36.34
CAD
TCC0440-025G
TCC0440-500G
Butyl cyanoacetate ≥98.0%
Butyl cyanoacetate
CAS Number: 5459-58-5
MDL Number: MFCD00001941
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 112
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: Cyanoacetic Acid Butyl Ester
Formula:
C₇H₁₁NO₂ Boiling Pt: 231 °C Melting Pt: –80 °C Density: 1.00 (20 °C) Flash Pt: 112 °C |
MDL Number:
MFCD00001941 CAS Number: 5459-58-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...