8-Chloroquinoline ≥98.0%
Supplier: TCI America
C0284-25G
C0284-5G
TCC0284-25GEA
761.56
CAD
TCC0284-25G
TCC0284-5G
8-Chloroquinoline ≥98.0%
8-Chloroquinoline
CAS Number: 611-33-6
MDL Number: MFCD00047618
Molecular Formula: C9H6ClN
Molecular Weight: 163.60
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 288
Specific Gravity (20/20): 1.28
Formula:
C₉H₆ClN Boiling Pt: 288 °C Melting Pt: –20 °C Density: 1.28 (20 °C) |
MDL Number:
MFCD00047618 CAS Number: 611-33-6 EINECS: 210-265-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...