5-Chloro-2-nitrobenzaldehyde ≥98.0%
Supplier: TCI America
C0217-5G
C0217-25G
TCC0217-005GEA
97.96
CAD
TCC0217-005G
TCC0217-25G
5-Chloro-2-nitrobenzaldehyde ≥98.0%
5-Chloro-2-nitrobenzaldehyde
CAS Number: 6628-86-0
MDL Number: MFCD00007289
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76
Formula:
C₇H₄ClNO₃ Melting Pt: 76 °C |
MDL Number:
MFCD00007289 CAS Number: 6628-86-0 UN: 3077 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...