1-Chlorohexadecane ≥98.0%

Supplier: TCI America

C0183-25ML C0183-500ML
TCC0183-025MLEA 46.8 CAD
TCC0183-025ML TCC0183-500ML
1-Chlorohexadecane ≥98.0%
1-Chlorohexadecane

CAS Number: 4860-03-1
MDL Number: MFCD00000959
Molecular Formula: C16H33Cl
Molecular Weight: 260.89
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 136
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Cetyl Chloride, Hexadecyl Chloride

Formula: C₁₆H₃₃Cl
Boiling Pt: 195 °C
Melting Pt: 7.8 °C
Density: 0.87 (20 °C)
Flash Pt: 136 °C
MDL Number: MFCD00000959
CAS Number: 4860-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b7be112e16421dadac05366b60090a512c6dd57&vid=f12c1ebd78e595cab705ce2f128c4437cc5521fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR