4-Chlorobenzoic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid

C0134-25G C0134-500G
TCC0134-025GEA 40.76 CAD
TCC0134-025G TCC0134-500G
4-Chlorobenzoic acid ≥99.0% (by GC, titration analysis)
4-Chlorobenzoic acid

CAS Number: 74-11-3
MDL Number: MFCD00002531
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 242
Lambda max.: 284 nm (EtOH)

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 242 °C
Density: 1.535 g/cm³ (20 °C)
Flash Pt: 238 °C
MDL Number: MFCD00000258
CAS Number: 74-11-3
Merck Index: 13,02144
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81a0465313a081e776a2a5d5a487f069e62d397e&vid=49da753ecdd5815fe0ad80a26b8e06d3352c2769&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR