Chalcone ≥98.0%
Supplier: TCI America
C0071-25G
C0071-100G
TCC0071-025GEA
61.48
CAD
TCC0071-025G
TCC0071-500G
TCC0071-100G
Chalcone ≥98.0%
Chalcone
CAS Number: 94-41-7
MDL Number: MFCD00003082
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 348
Melting point (°C): 57
Flash Point (°C): 113
Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2-Benzalacetophenone, 2-Benzylideneacetophenone, 1,3-Diphenyl-2-propen-1-one, Phenyl Styryl Ketone
Formula:
(C₆H₅)C(O)CHCH(C₆H₅) Boiling Pt: 348 °C Melting Pt: 57 °C Density: 1.0712 g/cm³ (62.4 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00003082 CAS Number: 94-41-7 Merck Index: 13,02045 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...