2-tert-Butylanthracene ≥98.0%
Supplier: TCI America
B3541-1G
B3541-5G
TCB3541-1GEA
173.8
CAD
TCB3541-1G
TCB3541-5G
2-tert-Butylanthracene ≥98.0%
2-tert-Butylanthracene
CAS Number: 18801-00-8
MDL Number: MFCD00003581
Molecular Formula: C18H18
Molecular Weight: 234.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 148
Formula:
C₁₈H₁₈ Boiling Pt: 363 °C (1013 hPa) Melting Pt: 148 °C |
MDL Number:
MFCD00003581 CAS Number: 18801-00-8 EINECS: 242-588-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...