5-Bromo-2-methoxypyrimidine ≥98.0%

Supplier: TCI America

B3521-5G B3521-1G
TCB3521-5GEA 262.28 CAD
TCB3521-5G TCB3521-1G
5-Bromo-2-methoxypyrimidine ≥98.0%
5-Bromo-2-methoxypyrimidine

CAS Number: 14001-66-2
MDL Number: MFCD04973351
Molecular Formula: C5H5BrN2O
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56

Formula: C₅H₅BrN₂O
Melting Pt: 56 °C
MDL Number: MFCD04973351
CAS Number: 14001-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=167f5f4dea35c7b3d407455b4928c246d3968721&vid=167f5f4dea35c7b3d407455b4928c246d3968721&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR