1-Butyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-Butyl-N-methylpyrrolidinium bromide
B3427-25G
B3427-5G
TCB3427-25GEA
234.79
CAD
TCB3427-25G
TCB3427-5G
1-Butyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidinium bromide
CAS Number: 93457-69-3
MDL Number: MFCD07784439
Molecular Formula: C9H20BrN
Molecular Weight: 222.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 214
Formula:
C₉H₂₀BrN MW: 222.17 g/mol Melting Pt: 214 °C |
MDL Number:
MFCD07784439 CAS Number: 93457-69-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...