1-Butyl-4-methylpyridinium chloride ≥99.0%
Supplier: TCI America
Synonyms:
1-Butyl-4-methylpyridinium Chloride
B3426-25G
B3426-5G
TCB3426-25GEA
551.42
CAD
TCB3426-25G
TCB3426-5G
1-Butyl-4-methylpyridinium chloride ≥99.0%
1-Butyl-4-methylpyridinium chloride
CAS Number: 112400-86-9
MDL Number: MFCD03095439
Molecular Formula: C10H16ClN
Molecular Weight: 185.70
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160
Formula:
C₁₀H₁₆ClN MW: 185.7 g/mol Melting Pt: 160 °C |
MDL Number:
MFCD03095439 CAS Number: 112400-86-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...