2-Bromo-4-fluorobenzyl alcohol ≥98.0%

Supplier: TCI America

B3417-5G B3417-25G
TCB3417-5GEA 137.46 CAD
TCB3417-5G TCB3417-25G
2-Bromo-4-fluorobenzyl alcohol ≥98.0%
2-Bromo-4-fluorobenzyl alcohol

CAS Number: 229027-89-8
MDL Number: MFCD00672925
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₇H₆BrFO
Melting Pt: 70 °C
MDL Number: MFCD00672925
CAS Number: 229027-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c775271123782cc0e180506064fd30a27bfafb&vid=51c775271123782cc0e180506064fd30a27bfafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR