4-Bromo-3,5-difluoroaniline ≥98.0%

Supplier: TCI America

B3416-5G B3416-25G
TCB3416-5GEA 99.54 CAD
TCB3416-5G TCB3416-25G
4-Bromo-3,5-difluoroaniline ≥98.0%
4-Bromo-3,5-difluoroaniline

CAS Number: 203302-95-8
MDL Number: MFCD00236593
Molecular Formula: C6H4BrF2N
Molecular Weight: 208.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 99

Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00236593
CAS Number: 203302-95-8
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a3da37a854604934195684970537d15527febdd&vid=5a3da37a854604934195684970537d15527febdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR