4-Bromo-2-methylthiophene ≥98.0%

Supplier: TCI America

B3385-25G B3385-5G
TCB3385-25GEA 399.74 CAD
TCB3385-25G TCB3385-5G
4-Bromo-2-methylthiophene ≥98.0%
4-Bromo-2-methylthiophene

CAS Number: 29421-92-9
MDL Number: MFCD04116367
Molecular Formula: C5H5BrS
Molecular Weight: 177.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): 68
Specific Gravity (20/20): 1.58

Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 62 °C
Density: 1.58 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD04116367
CAS Number: 29421-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd678eb7e1efb123c30587aaa41a206d97779f9c&vid=bd678eb7e1efb123c30587aaa41a206d97779f9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR